missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARG2 Antibody (OTI3G5), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70205APC
This item is not returnable.
View return policy
Description
ARG2 Monoclonal antibody specifically detects ARG2 in Human, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| ARG2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Canine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI3G5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| arginase 2, arginase, type II, arginase-2, mitochondrial, EC 3.5.3, EC 3.5.3.1, kidney arginase, Kidney-type arginase, L-arginine amidinohydrolase, L-arginine ureahydrolase, nonhepatic arginase, Non-hepatic arginase, Type II arginase | |
| Full length human recombinant protein of human ARG2(NP_001163) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 384 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction