missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody (ARG1/1126), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47999UV
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Monoclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Arginase 1/ARG1/liver Arginase | |
| Monoclonal | |
| DyLight 350 | |
| ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase | |
| Recombinant fragment (around aa11-97) of human Arginase 1/ARG1/liver Arginase protein(exact sequence is proprietary) (Uniprot: P05089) | |
| 0.1 mL | |
| Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction | |
| 383 | |
| Store at 4°C in the dark. | |
| IgG3 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| ARG1/1126 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction