missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody (ARG1/8869R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24201JF646
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Monoclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Arginase 1/ARG1/liver Arginase | |
| Monoclonal | |
| Janelia Fluor 646 | |
| ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase | |
| Recombinant human Arginase 1/ARG1/liver Arginase protein fragment (aa1-200) (Exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction | |
| 383 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| ARG1/8869R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction