missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody (OTI4E6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70204F
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Arginase 1/ARG1/liver Arginase | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ARG1 | |
| Full length recombinant protein of human Arginase 1/ARG1/liver Arginase (NP_000036) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI4E6 | |
| Flow Cytometry, Immunohistochemistry | |
| arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 383 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction