missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase-1 (ARG1) Mouse anti-Mouse, Rat, Clone: UMAB288, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies UM870178
This item is not returnable.
View return policy
Description
Arginase-1 (ARG1) Monoclonal antibody specifically detects Arginase-1 (ARG1) in Mouse, Rat samples. It is validated for Immunohistochemistry, Western BlotSpecifications
| Arginase-1 (ARG1) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07824, Q61176 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11846, 29221 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| UMAB288 | |
| Unconjugated | |
| ARG1 | |
| AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase | |
| Full length human recombinant protein of human ARG1 (NP_000036) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction