Learn More
Argininosuccinate Lyase Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99244APC
Product Code. 18688425
  • € 407.00 / 0.10mL
Description
Argininosuccinate Lyase Polyclonal antibody specifically detects Argininosuccinate Lyase in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry (Paraffin)
APC
Store at 4C in the dark.