missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Lyase Antibody (OTI3D4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70213AF750
This item is not returnable.
View return policy
Description
Argininosuccinate Lyase Monoclonal specifically detects Argininosuccinate Lyase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Argininosuccinate Lyase | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ASL | |
| Full length recombinant protein of human Argininosuccinate Lyase (NP_001020114) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI3D4 | |
| Western Blot | |
| argininosuccinase, argininosuccinate lyase, Arginosuccinase, ASAL, EC 4.3.2.1 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 435.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction