missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Synthase Antibody (2C10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-37518
This item is not returnable.
View return policy
Description
Argininosuccinate Synthase Monoclonal specifically detects Argininosuccinate Synthase in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Argininosuccinate Synthase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5 | |
| Mouse | |
| 47 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Monkey | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2C10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ASS1 | |
| Purified recombinant fragment of human ASS1 expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 445 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction