missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginyl tRNA synthetase Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97635H
This item is not returnable.
View return policy
Description
Arginyl tRNA synthetase Polyclonal specifically detects Arginyl tRNA synthetase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Arginyl tRNA synthetase | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| arginine tRNA ligase 1, cytoplasmic, Arginine--tRNA ligase, arginyl-tRNA synthetase, arginyl-tRNA synthetase, cytoplasmic, ArgRS, DALRD1, EC 6.1.1.19, MGC8641 | |
| Produced in rabbits immunized with E. coli-derived Human Arginyl tRNA synthetase fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 5917 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction