missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Arginyl tRNA synthetase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30609
This item is not returnable.
View return policy
Description
Arginyl tRNA synthetase Polyclonal antibody specifically detects Arginyl tRNA synthetase in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Arginyl tRNA synthetase | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5917 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| arginine tRNA ligase 1, cytoplasmic, Arginine--tRNA ligase, arginyl-tRNA synthetase, arginyl-tRNA synthetase, cytoplasmic, ArgRS, DALRD1, EC 6.1.1.19, MGC8641 | |
| Between 1 and 50 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction