missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginyl tRNA synthetase Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-46148
This item is not returnable.
View return policy
Description
Arginyl tRNA synthetase Polyclonal specifically detects Arginyl tRNA synthetase in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Arginyl tRNA synthetase | |
| Polyclonal | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_002878.2 | |
| RARS1 | |
| Peptide with sequence C-SLQAERNKPTKN corresponding to internal region according to NP_002878.2. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human. Expected from sequence similarity: Mouse and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| arginine tRNA ligase 1, cytoplasmic, Arginine--tRNA ligase, arginyl-tRNA synthetase, arginyl-tRNA synthetase, cytoplasmic, ArgRS, DALRD1, EC 6.1.1.19, MGC8641 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5917 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction