missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARGLU1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566041
This item is not returnable.
View return policy
Description
Immunogen sequence: RSRSTNTAVSR RERDRERASS PPDRIDIFGR TVSKRSSLDE KQKR Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
ARGLU1 belongs to the UPF0430 family. There are two named isoforms.
Specifications
| ARGLU1 | |
| Polyclonal | |
| Unconjugated | |
| ARGLU1 | |
| 9430010O03Rik; arginine and glutamate rich 1; arginine and glutamate-rich protein 1; arginine and glutamate-rich protein 1-A; LOW QUALITY PROTEIN: arginine and glutamate-rich protein 1; ARGLU1; C130008N12; LOW QUALITY PROTEIN: arginine and glutamate-rich protein 1; RGD1310061; Unknown (protein for MGC:134421) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWB6 | |
| ARGLU1 | |
| Recombinant Human ARGLU1. Recombinant protein control fragment (Product #RP-105053). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction