missing translation for 'onlineSavingsMsg'
Learn More

ARH Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30511368 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30511368 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511368 Supplier Novus Biologicals Supplier No. NBP338382CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARH Polyclonal antibody specifically detects ARH in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARH
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 59-308 of human ARH (NP_056442.2).,, Sequence:, EQPKGEELSAAAIKRIVATAKASGKKLQKVTLKVSPRGIILTDNLTNQLIENVSIYRISYCTADKMHDKVFAYIAQSQHNQSLECHAFLCTKRKMAQAVTLTVAQAFKVAFEFWQVSKEEKEKRDKASQEGGDVLGARQDCTPSLKSLVATGNLLDLEETAKAPLSTVSANTTNMDEVPRPQALSGSSVVWELDDGLDEAFSRLAQSRTNPQVLDTGLTAQDMHYAQCLSPVDWDKPDSSGTEQDDLFSF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 26119
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.