missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARH Antibody (OTI7E11), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71790AF594
This item is not returnable.
View return policy
Description
ARH Monoclonal specifically detects ARH in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ARH | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LDLRAP1 | |
| Full length recombinant protein of human ARH (NP_056442) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI7E11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 26119.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction