missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP12 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809263
This item is not returnable.
View return policy
Description
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.Specifications
| ARHGAP12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IWW6 | |
| ARHGAP12 | |
| Human recombit protein fragment corresponding to amino acids 286-452 of human ARHGAP12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4A3 | |
| Unconjugated | |
| ARHGAP12 | |
| Rho GTPase activating protein 12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 94134 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction