missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGAP23 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16640555 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16640555 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16640555 Supplier Invitrogen™ Supplier No. PA5104103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHG23.

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP23
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP23
Gene Accession No. Q69ZH9, Q9P227
Gene Alias ARHGAP23; KIAA1501; Rho GTPase activating protein 23; rho GTPase-activating protein 23; Rho-type GTPase-activating protein 23
Gene Symbols ARHGAP23
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP23(Accession Q9P227), corresponding to amino acid residues P495-S545.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303501, 57636, 58996
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.