missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557238
This item is not returnable.
View return policy
Description
Immunogen sequence: IQRVMTMMIR DHEVLFPKSK DIPLSPPAQK NDPKKAPVAR SSVGWDATED LRISRTDSFS SMTSDSDTTS PTGQQPSDAF PEDSSKVPRE K Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.
ARHGAP25 encode negative regulators of Rho GTPases, which are implicated in actin remodeling, cell polarity, and cell migration.
Specifications
| ARHGAP25 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP25 | |
| A130039I20Rik; ARHGAP25; epididymis secretory protein Li 308; HEL-S-308; KAIA0053; KIAA0053; RGD1562105; Rho GTPase activating protein 25; rho GTPase-activating protein 25; Rho-type GTPase-activating protein 25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9938 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P42331 | |
| ARHGAP25 | |
| Recombinant protein corresponding to Human ARHGAP25. Recombinant protein control fragment (Product #RP-95003). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction