Learn More
Invitrogen™ ARHGAP25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589350
Description
Immunogen sequence: MSLGQSACLF LSIARSRSVM TGEQMAAFHP SSTPNPLERP IKMGWLKKQR SIVKNWQQRY FVLRAQQLYY YKDEEDTKPQ GCMYLPGCTI KEIATNPEEA GKFVFEIIPA SWDQNRMGQD SYVLMASSQA EMEEWVKFLR RVAGTPCGAV FGQRLDETVA YEQKFGPHLV PILVEKCAEF ILEHGRNEEG IFRLPGQDNL VKQLRDAFDA GERPSFDRDT DVHTVASLLK LYLRDLPEPV VPWSQYEGFL LCGQLTNADE AKAQQELMKQ LSILPRDNYS LLSYICRFLH EIQLNCAVNK MSVDNLATVI GVNLIRSKVE DPAVIMRGTP QIQRVMTMMI RDHEVLFPKS KDIPLSPPAQ KNDPKKAPVA; Positive Samples: Jurkat, A-549, Mouse lung, Rat spleen.
ARHGAP25 encode negative regulators of Rho GTPases, which are implicated in actin remodeling, cell polarity, and cell migration.
Specifications
| ARHGAP25 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP25 | |
| A130039I20Rik; ARHGAP25; epididymis secretory protein Li 308; HEL-S-308; KAIA0053; KIAA0053; RGD1562105; Rho GTPase activating protein 25; rho GTPase-activating protein 25; Rho-type GTPase-activating protein 25 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232201, 500246, 9938 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42331, Q8BYW1 | |
| ARHGAP25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-370 of human ARHGAP25 (NP_0011597481). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.