missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGAP27 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15968494
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15968494 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15968494 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA565759

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PRSIHKSSQDG DTPAQASPPE EKVPAELDEV GSWEEVSPAT AAVRTKTLDK AGVLHRTKTA DKGKRLRKEH WSAS Highest antigen sequence identity to the following orthologs - mouse 61%, rat 61%.

Rho GTPase-activating protein which may be involved in clathrin-mediated endocytosis. GTPase activators for the Rho-type GTPases act by converting them to an inactive GDP-bound state. Has activity toward CDC42 and RAC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP27
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP27
Gene Accession No. Q6ZUM4
Gene Alias ARHGAP27; CAMGAP1; CIN85-associated multi-domain-containing Rho GTPase-activating protein 1; PP905; Rho GTPase activating protein 27; rho GTPase-activating protein 27; Rho-type GTPase-activating protein 27; SH3 domain containing 20; SH3 domain-containing protein 20; SH3D20; SH3P20
Gene Symbols ARHGAP27
Host Species Rabbit
Immunogen Recombinant Human ARHGAP27. Recombinant protein control fragment (Product #RP-104960).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201176
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.