missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP4 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97914B
This item is not returnable.
View return policy
Description
ARHGAP4 Polyclonal antibody specifically detects ARHGAP4 in Human samples. It is validated for Western Blot
Specifications
| ARHGAP4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 393 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| RGC1, Rho GTPase activating protein 4 | |
| Produced in rabbits immunized with E. coli-derived Human ARHGAP4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction