missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP5 Monoclonal Antibody (OTI9H2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527152
This item is not returnable.
View return policy
Description
ARHGAP5 Monoclonal Antibody for Western Blot
Rho GTPase activating protein 5 negatively regulates RHO GTPases, a family which may mediate cytoskeleton changes by stimulating the hydrolysis of bound GTP. Two transcript variants encoding different isoforms have been found for this gene. GTPase-activating protein for Rho family members. May play a role in the reduction of the p21rasGTPase-activating potential of RASA1/p120GAP.
Specifications
| ARHGAP5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13017 | |
| ARHGAP5 | |
| Human recombinant protein fragment corresponding to amino acids 948-1261 of ARHGAP5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI9H2 | |
| Unconjugated | |
| ARHGAP5 | |
| ARHGAP5; GFI2; growth factor independent 2; LRRGT00098; p100 RasGAP-associated p105 protein; p105 RhoGAP; p190B; p190-B; p190BRhoGAP; Rho GTPase activating protein 5; rho GTPase-activating protein 5; Rho GTPase-activating protein 5-like protein; RHOGAP5; rho-type GTPase-activating protein 5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 394 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction