missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGDIG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106722
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARHGDIG.
The Ras superfamily of small GTP-binding proteins are critical mediators of diverse cell signaling pathways, including those leading to proliferation, cytoskeletal organization and secretion. The counter-conversion of the active GTPbound form of these proteins to their inactive GDP-bound form is influenced by two types of regulatory proteins: those that alter the intrinsic GTPase activity of the GTP-binding proteins and those that alter the rate of GDP/GTP exchange. Guanine nucleotide-releasing factors (GRFs) increase the GDP dissociation rate, while GDP-dissociation inhibitors (GDIs) decrease the dissociation rate. The Rho GDI subfamily is composed of Rho GDIalpha, Ly-GDI (also known as Rho GDIbeta and previously known as GDI/D4) and Rho GDIgamma. The Rho GDI proteins interact with and have varying affinities for several Ras-like GTP binding proteins, including Rho A, Rho B, Rac and Cdc42. Ly-GDI is expressed only in hematopoietic cells, predominantly in B and T lymphocyte cell lines.
Specifications
| ARHGDIG | |
| Polyclonal | |
| Unconjugated | |
| ARHGDIG | |
| Arhgdig; Gdi5; guanosine diphosphate (GDP) dissociation inhibitor 5, Rho associated; rho GDI 3; Rho GDIgamma; Rho GDP dissociation inhibitor (GDI) gamma; Rho GDP dissociation inhibitor gamma; rho GDP-dissociation inhibitor 3; RhoGDI gamma; Rho-GDI gamma; Rho-GDI2; Rho-GDI-3; RHOGDI-3; Rho-GDIgamma; RIP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 398 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99819 | |
| ARHGDIG | |
| A synthesized peptide derived from human ARHGDIG(Accession Q99819), corresponding to amino acid residues R60-D110. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction