missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46831AF405
This item is not returnable.
View return policy
Description
ARHGEF1 Polyclonal antibody specifically detects ARHGEF1 in Human samples. It is validated for Western Blot
Specifications
| ARHGEF1 | |
| Polyclonal | |
| Western Blot | |
| 115 kDa guanine nucleotide exchange factor, 115-kD protein, GEF1, LBCL2LSC, Lsc homolog, p115RhoGEF, P115-RHOGEF, Rho guanine nucleotide exchange factor (GEF) 1, rho guanine nucleotide exchange factor 1, Sub1.5 | |
| The immunogen for this product maps to a region between residue 775 and 825 of human 115 kDa Rho guanine nucleotide exchange factor using the numbering given in entry NP_004697.2 (GeneID 9138). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 9138 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction