missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF1 Polyclonal Antibody

Description
Immunogen sequence: PLLSSSENGN GGRETSPADA RTERILSDLL PFCRPGPEGQ LAATALRKVL SLKQLLFPAE EDNGAGPPRD GDGVPGGGPL SPARTQEIQE NLLSLEETMK QLEELEEEFC RLRPLLSQLG GNSVPQPGCT; Positive Samples: HeLa, Jurkat, LO2; Cellular Location: Cytoplasm, Membrane.
The Ras superfamily of GTPases, which can be subdivided into the Ras, Rho/Rac, Sar, Rab, ARF and Ran subfamilies, controls multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. The Ras superfamily of GTPases function as regulated switches that toggle between a biologically active GTP-bound and an inactive GDP-bound form. This activation is catalyzed by guanine nucleotide exchange factors (GEFs). The Dbl-related proteins are a large family of structurally related molecules that have a common ability to catalyze GEF activity for specific members of the Ras family. Dbl-related proteins include FGD1, Lsc, RhoGEF p115, Lfc, Lbc and Brx. Lsc, Lbc and Lfc share sequence homology and show exchange activity toward Rho family GTPases. RhoGEF p115 catalyzes GEF activity for Rho but not Rac, Cdc42 or Ras GTPases.
Specifications
Specifications
| Antigen | ARHGEF1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Arhgef1 |
| Gene Accession No. | Q92888, Q9Z1I6 |
| Gene Alias | 115 kDa guanine nucleotide exchange factor; 115-kD protein; ARHGEF1; GEF1; LBCL2; Lbc's second cousin; Lsc; Lsc homolog; lymphoid blast crisis like 2; lymphoid blast crisis-like 2; p115RhoGEF; p115-RhoGEF; Rho guanine nucleotide exchange factor (GEF) 1; Rho guanine nucleotide exchange factor 1; Sub1.5 |
| Gene Symbols | Arhgef1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?