missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF18 Monoclonal Antibody (OTI6D10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527295
This item is not returnable.
View return policy
Description
ARHGEF18 Monoclonal Antibody for Western Blot, IHC (P)
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ARHGEF18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZSZ5 | |
| Arhgef18 | |
| Human recombinant protein fragment corresponding to amino acids 118-215 of ARHGEF18 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6D10 | |
| Unconjugated | |
| Arhgef18 | |
| 114 kDa Rho-specific guanine nucleotide exchange factor; AI467246; Arhgef18; D030053O22Rik; KIAA0521; p114RhoGEF; P114-RhoGEF; p114-Rho-GEF; rho guanine nucleotide exchange factor 18; rho/rac guanine nucleotide exchange factor (GEF) 18; Rho/Rac guanine nucleotide exchange factor 18; Rho-specific guanine nucleotide exchange factor p114; SA-RhoGEF; Septin-associated RhoGEF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23370 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction