missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID1A Antibody (ARID1A/7735) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-27036JF669
This item is not returnable.
View return policy
Description
ARID1A Monoclonal antibody specifically detects ARID1A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ARID1A | |
| Monoclonal | |
| Janelia Fluor 669 | |
| ARID domain-containing protein 1A, AT rich interactive domain 1A (SWI- like), AT rich interactive domain 1A (SWI-like), AT-rich interactive domain-containing protein 1A, B120SWI-like protein, BAF250a, BAF250SWI/SNF complex protein p270, BM029, brain protein 120, BRG1-associated factor 250, BRG1-associated factor 250a, C10rf4, C1orf4SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatinsubfamily F member 1, chromatin remodeling factor p250, hELD, hOSA1, matrix associated, actin dependent regulator of chromatin, Osa homolog 1, OSA1, OSA1 nuclear protein, P270, SMARCF1, subfamily f, member 1 | |
| Recombinant fragment (around aa1900-2000) of human ARID1A protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Signal Transduction | |
| 8289 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| ARID1A/7735 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction