missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID1A Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98979FR
This item is not returnable.
View return policy
Description
ARID1A Polyclonal antibody specifically detects ARID1A in Human samples. It is validated for Western Blot
Specifications
| ARID1A | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| ARID domain-containing protein 1A, AT rich interactive domain 1A (SWI- like), AT rich interactive domain 1A (SWI-like), AT-rich interactive domain-containing protein 1A, B120SWI-like protein, BAF250a, BAF250SWI/SNF complex protein p270, BM029, brain protein 120, BRG1-associated factor 250, BRG1-associated factor 250a, C10rf4, C1orf4SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatinsubfamily F member 1, chromatin remodeling factor p250, hELD, hOSA1, matrix associated, actin dependent regulator of chromatin, Osa homolog 1, OSA1, OSA1 nuclear protein, P270, SMARCF1, subfamily f, member 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ARID1A | |
| 0.1 mL | |
| Signal Transduction | |
| 8289 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction