missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID5A Antibody (18112), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43745
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
ARID5A Monoclonal specifically detects ARID5A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| ARID5A | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| ARID5A | |
| Recombinant protein encompassing a sequence within the center region of human ARID5A. The exact sequence is proprietary. | |
| Protein A purified | |
| RUO | |
| 10865 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 18112 | |
| Western Blot 1:500-1:10000, Flow Cytometry 1:20-1:200, Immunohistochemistry, Immunohistochemistry-Frozen 1:100-1:1000 | |
| ARID domain-containing protein 5A, AT rich interactive domain 5A (MRF1-like), AT-rich interactive domain-containing protein 5A, RFVG5814 | |
| Mouse | |
| 64 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction