missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARID5A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA532020

Product Code. 13486587

  • € 532.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 3xFlag-human ARID5A-transfected 293T. Predicted reactivity: Mouse (83%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARID5A
Polyclonal
Unconjugated
ARID5A
ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14
Rabbit
Antigen affinity chromatography
RUO
10865
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.55 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q03989
ARID5A
Recombinant protein encompassing a sequence within the center region of human ARID5A. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.