missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARID5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553270
This item is not returnable.
View return policy
Description
Immunogen sequence: TSKYPSRDMY RESENSSFPS HRHQEKLHVN YLTSLHLQDK KSAAAEAPTD DQPTDLSLPK NPHKPTGKVL GLAHSTTGPQ ESKGISQFQV LGSQSRDCHP KACRVSPMTM SGPKKYPESL SRSGKPHHVR LENFRKME Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 24%.
ARID5B encodes a member of the AT-rich interaction domain (ARID) family of DNA binding proteins. The encoded protein forms a histone H3K9Me2 demethylase complex with PHD finger protein 2 and regulates the transcription of target genes involved in adipogenesis and liver development. ARID5B also plays a role in cell growth and differentiation of B-lymphocyte progenitors, and single nucleotide polymorphisms in this gene are associated with acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| ARID5B | |
| Polyclonal | |
| Unconjugated | |
| ARID5B | |
| ARID domain-containing protein 5B; ARID5B; AT rich interactive domain 5B (MRF1-like); AT-rich interaction domain 5B; AT-rich interactive domain 5B (MRF1-like); AT-rich interactive domain-containing protein 5B; DESRT; Modulator recognition factor 2; modulator recognition factor 2 (MRF2); MRF1-like protein; MRF2; MRF-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14865 | |
| ARID5B | |
| Recombinant protein corresponding to Human MRF2. Recombinant protein control fragment (Product #RP-90906). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction