missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARIH2 Antibody (OTI3F9), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71600IR
This item is not returnable.
View return policy
Description
ARIH2 Monoclonal specifically detects ARIH2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ARIH2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARIH2 | |
| Recombinant protein fragment corresponding to amino acids 201-493 of human ARIH2 (NP_006312) produced in E.coli. | |
| 0.1 mL | |
| 10425.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3F9 | |
| Western Blot | |
| ARI-2, ariadne (Drosophila) homolog 2, ariadne homolog 2 (Drosophila), E3 ubiquitin-protein ligase ARIH2, EC 6.3.2.-, FLJ10938, FLJ33921, Protein ariadne-2 homolog, Triad1 protein, TRIAD1ARI2all-trans retinoic acid inducible RING finger | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction