Learn More
Sartorius Arium™ Advance L Pre-filter Tubing Set
Arium™ Advance L Filter Tubing Set for direct and uncomplicated connection of single or dual filter housings.
Brand: Sartorius H2O-AHSL-1
237.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The Arium™ Advance L filter tubing set is used for direct and uncomplicated connection of single or dual filter housings used as pre-treatment for feed water with an Arium™ pure water system.
Specifications
EPDM braided blue poly tubing with 0.75 in. stainless steel female adapter | |
Reverse Osmosis (Type 3) | |
1.50 m |
Potable Tap Water | |
Tubing |