Learn More
Sartorius™ arium™ bagtanks
Shop All Sartorius Products
Click to view available options
Capacity (Metric):
100 L
Unit Size:
Each
Description
- Integrated venting filter with check valve securely protects against CO2 contamination
- Optional rollers provide flexibility
Specifications
Specifications
| Capacity (Metric) | 100 L |
| For Use With (Application) | Pure water is securely protected from secondary contamination |
| Material | Stainless Steel, Plastic |
| Sterility | Sterile |