missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13A Polyclonal Antibody

Brand: Thermo Scientific PA5114280
This item is not returnable.
View return policy
Description
Specifications
| ARL13A | |
| Polyclonal | |
| Unconjugated | |
| ARL13A | |
| ADP ribosylation factor like GTPase 13A; ADP-ribosylation factor like GTPase 13A; ADP-ribosylation factor-like 13; ADP-ribosylation factor-like 13A; ADP-ribosylation factor-like protein 13A; ARL13; ARL13A; dJ341D10.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 392509 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5H913 | |
| ARL13A | |
| Synthetic peptide directed towards the middle region of Human ARL13A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction