missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL15 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92298-0.02ml
This item is not returnable.
View return policy
Description
ARL15 Polyclonal antibody specifically detects ARL15 in Mouse samples. It is validated for Western Blot
Specifications
| ARL15 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ADP-ribosylation factor related protein 2, ADP-ribosylation factor-like 15, ADP-ribosylation factor-like protein 15, ADP-ribosylation factor-related protein 2, ARF-related protein 2, ARFRP2, FLJ20051 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-204 of human ARL15 (NP_061960.1). MSDLRITEAFLYMDYLCFRALCCKGPPPARPEYDLVCIGLTGSGKTSLLSKLCSESPDNVVSTTGFSIKAVPFQNAILNVKELGGADNIRKYWSRYYQGSQGVIFVLDSASSEDDLEAARNELHSALQHPQLCTLPFLILANHQDKPAARSVQEIKKYFELEPLARGKRWILQPCSLDDMDALKDSFSQLINLLEEKDHEAVRM | |
| 0.02 mL | |
| GPCR, Signal Transduction | |
| 54622 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction