missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL15 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77052IR
This item is not returnable.
View return policy
Description
ARL15 Polyclonal antibody specifically detects ARL15 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ARL15 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| ADP-ribosylation factor related protein 2, ADP-ribosylation factor-like 15, ADP-ribosylation factor-like protein 15, ADP-ribosylation factor-related protein 2, ARF-related protein 2, ARFRP2, FLJ20051 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human ARL15. The immunogen is located within amino acids 130 - 180 of ARL15. Amino Acid Squence: NHQDKPAARSVQEIKKY | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 54622 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction