missing translation for 'onlineSavingsMsg'
Learn More

ARL17A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15799151 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15799151 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15799151 Supplier Thermo Scientific Supplier No. PA561339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARL17A Polyclonal Antibody

ARL17A Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL17A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL17A
Gene Accession No. Q8IVW1
Gene Alias ADP ribosylation factor like GTPase 17A; ADP-ribosylation factor 7; ADP-ribosylation factor 7 variant; ADP-ribosylation factor-like 17-like; ADP-ribosylation factor-like protein 17; ARF1P2; ARF1P2 ARL17A; ARL17A; ARL17B; ARL17P1; PRO2667
Gene Symbols ARL17A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL17A. Recombinant protein control fragment (Product #RP-104408).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.