missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527091
This item is not returnable.
View return policy
Description
Recommended positive controls: Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Xenopus laevis (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a small GTP-binding protein of the RAS superfamily which functions as an ADP-ribosylation factor (ARF). The encoded protein is one of a functionally distinct group of ARF-like genes.
Specifications
| ARL2 | |
| Polyclonal | |
| Unconjugated | |
| ARL2 | |
| 2610009M23Rik; ADP ribosylation factor like GTPase 2; ADP-ribosylation factor like GTPase 2; ADP-ribosylation factor-like 2; ADP-ribosylation factor-like protein 2; ADP-ribosylation-like 2; AI115441; ARFL2; arf-like protein 2; Arl184; Arl2; AW553335 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 402, 56327, 65142 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O08697, P36404, Q9D0J4 | |
| ARL2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ARL2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction