missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL2BP Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00281APC
This item is not returnable.
View return policy
Description
ARL2BP Polyclonal antibody specifically detects ARL2BP in Human samples. It is validated for ELISA
Specifications
| ARL2BP | |
| Polyclonal | |
| ELISA | |
| ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2 | |
| Produced in rabbits immunized with purified, recombinant Human ARL2BP (Uniprot#: Q9Y2Y0-1; Met1-His163). | |
| 0.1 mL | |
| Signal Transduction | |
| 23568 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction