missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL2BP Antibody (OTI1A10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71743H
This item is not returnable.
View return policy
Description
ARL2BP Monoclonal specifically detects ARL2BP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ARL2BP | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ARL2BP | |
| Full length recombinant protein of human ARL2BP (NP_036238) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1A10 | |
| Western Blot, Immunohistochemistry | |
| ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 23568.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction