missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL3 Antibody (OTI1C6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70206H
This item is not returnable.
View return policy
Description
ARL3 Monoclonal specifically detects ARL3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ARL3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ARL3 | |
| Recombinant protein fragment corresponding to amino acids 2-182 of human ARL3 (NP_004302) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1C6 | |
| Western Blot, Immunohistochemistry | |
| ADP-ribosylation factor-like 3, ARFL3ADP-ribosylation factor-like protein 3, ARF-like 3 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 403.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction