Learn More
ARL3 Monoclonal Antibody (OTI3E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803089S
Encompass_Preferred
 View more versions of this product
Product Code. 17571983
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ARL3 Monoclonal antibody specifically detects ARL3 in Human samples. It is validated for Western Blot
Specifications
Specifications
ARL3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P36405
Mouse
Affinity chromatography
RUO
403
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3E1
Unconjugated
Arl3
ADP ribosylation factor like GTPase 3; ADP-ribosylation factor like GTPase 3; ADP-ribosylation factor-like 3; ADP-ribosylation factor-like protein 3; ADP-ribosylation-like 3; ARD3; ARFL3; ARF-like 3; Arf-like protein 3; Arl3
Human recombinant protein fragment corresponding to amino acids 2-182 of human ARL3 produced in E.coli.
30 μL