missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL3 Monoclonal Antibody (OTI7E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803085S
This item is not returnable.
View return policy
Description
ARL3 Monoclonal antibody specifically detects ARL3 in Human samples. It is validated for Western BlotSpecifications
| ARL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHR5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29115 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI7E5 | |
| Unconjugated | |
| Sap30bp | |
| ADP ribosylation factor like GTPase 3; ADP-ribosylation factor like GTPase 3; ADP-ribosylation factor-like 3; ADP-ribosylation factor-like protein 3; ADP-ribosylation-like 3; ARD3; ARFL3; ARF-like 3; Arf-like protein 3; Arl3 | |
| Human recombinant protein fragment corresponding to amino acids 1-260 of human SAP30BP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction