missing translation for 'onlineSavingsMsg'
Learn More

ARL6IP4 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 209.00 - € 463.00

Specifications

Antigen ARL6IP4
Dilution Western Blot 1:1000 - 1:2000, ELISA
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18693882
View Documents
Novus Biologicals
NBP2-92711-0.02ml
0.02 mL
€ 209.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18680552
View Documents
Novus Biologicals
NBP2-92711-0.1ml
0.1 mL
€ 463.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ARL6IP4 Polyclonal antibody specifically detects ARL6IP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications

Specifications

ARL6IP4
Western Blot, ELISA
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Signal Transduction
PBS with 50% glycerol, pH7.3.
51329
IgG
Affinity purified
Western Blot 1:1000 - 1:2000, ELISA
Polyclonal
Purified
RUO
Human, Mouse, Rat
ADP-ribosylation-like factor 6 interacting protein 4, Aip-4, ARL-6-interacting protein 4, HSP-975, HSVI binding protein, HSVI-binding protein, MGC814, SFRS20, splicing factor, arginine/serine-rich 20, splicing regulator SRrp38, SR-15, SR-25ADP-ribosylation factor-like protein 6-interacting protein 4, SRp25 nuclear protein, SRp25aip-4, SRrp37
Recombinant fusion protein containing a sequence corresponding to amino acids 313-402 of human ARL6IP4 (NP_057722.3). SAGEEEDGPVLTDEQKSRIQAMKPMTKEEWDARQSIIRKVVDPETGRTRLIKGDGEVLEEIVTKERHREINKQATRGDCLAFQMRAGLLP
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.