missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144869
This item is not returnable.
View return policy
Description
Immunogen sequence: KQDLEAAYHI TDIHEALALS EVGNDRKMFL FGTYLTKNGS EIPSTMQDAK DLIA Antigen sequence identity: Mouse 91%; Rat 83%.
ARL9 is a member of the small GTPase protein family with a high degree of similarity to ARF proteins of the RAS superfamily.
Specifications
| ARL9 | |
| Polyclonal | |
| Unconjugated | |
| ARL9 | |
| 1700110A09Rik; ADP ribosylation factor like GTPase 9; ADP-ribosylation factor like GTPase 9; ADP-ribosylation factor-like 9; ADP-ribosylation factor-like protein 9; ARL9; Unknown (protein for MGC:153259); zgc:153259 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 132946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6T311 | |
| ARL9 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 127-180. Recombinant protein control fragment (Product #RP-104205). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction