missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC1 Antibody (OTI1D10), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71989JF549
This item is not returnable.
View return policy
Description
ARMC1 Monoclonal specifically detects ARMC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ARMC1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARMC1 | |
| Full length recombinant protein of human ARMC1 (NP_060590) produced in HEK293T cells. | |
| 0.1 mL | |
| 55156.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Arcp, armadillo repeat containing 1, armadillo repeat-containing protein 1, FLJ10511 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction