missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37916-20ul
This item is not returnable.
View return policy
Description
ARMC10 Polyclonal antibody specifically detects ARMC10 in Human samples. It is validated for ELISA,Western Blot
Specifications
| ARMC10 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| armadillo repeat containing 10, armadillo repeat-containing protein 10, FLJ37850, MGC3195, specific splicing variant involved in hepatocarcinogenesis, Splicing variant involved in hepatocarcinogenesis protein, SVHPNAS112 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 169-308 of human ARMC10 (NP_001154481.1).,, Sequence:, TNMTVTNDHQHMLHSYITDLFQVLLTGNGNTKVQVLKLLLNLSENPAMTEGLLRAQVDSSFLSLYDSHVAKEILLRVLTLFQNIKNCLKIEGHLAVQPTFTEGSLFFLLHGEECAQKIRALVDHHDAEVKEKVVTIIPKI | |
| 20 μL | |
| Cancer | |
| 83787 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction