Få mere at vide
Få mere at vide
ARMC3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28418-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The ARMC3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARMC3. This antibody recognizes human, mouse antigen. The ARMC3 antibody has been validated for the following applications: IP, ELISA analysis.
Tekniske data
| ARMC3 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunoprecipitation, ELISA | |
| armadillo repeat containing 3, ARMC3, Beta catenin like protein, Cancer/testis antigen 81, CT81, FLJ25845, FLJ32827, KU CT 1 | |
| 20 μL | |
| 219681 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion