Learn More
ARMC3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28418-1-AP-150UL
 View more versions of this product
Product Code. 18075135
  • € 410.00 / 150µL
Description
The ARMC3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARMC3. This antibody recognizes human, mouse antigen. The ARMC3 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
Specifications
ARMC3
400 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunoprecipitation, ELISA
armadillo repeat containing 3, ARMC3, Beta catenin like protein, Cancer/testis antigen 81, CT81, FLJ25845, FLJ32827, KU CT 1
150 μL
219681
Store at -20°C. Stable for one year after shipment.