Learn More
ARMC3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28418-1-AP-150UL
This item is not returnable.
View return policy
Description
The ARMC3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARMC3. This antibody recognizes human, mouse antigen. The ARMC3 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
| ARMC3 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunoprecipitation, ELISA | |
| armadillo repeat containing 3, ARMC3, Beta catenin like protein, Cancer/testis antigen 81, CT81, FLJ25845, FLJ32827, KU CT 1 | |
| 150 μL | |
| 219681 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |