missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARMC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106605
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARMC6.
The function of this gene's protein product has not been determined. A related protein in mouse suggests that this protein has a conserved function.
Specifications
| ARMC6 | |
| Polyclonal | |
| Unconjugated | |
| Armc6 | |
| 2410153K17Rik; armadillo repeat containing 6; armadillo repeat-containing protein 6; Armc6; AW554412; R30923_1; RGD1306691 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 306352, 76813, 93436 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6NXE6, Q8BNU0 | |
| Armc6 | |
| A synthesized peptide derived from human ARMC6(Accession Q6NXE6), corresponding to amino acid residues H444-G494. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction